Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022590.2 | NP_072112.2 | 5 | 561 | ||
GenBank mRNA | AB070849.1 | - | 5 | 556 | ||
AB070850.1 | - | 5 | 556 | |||
AB070976.1 | - | 5 | 556 | |||
U29881.1 | - | 5 | 561 |