Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022602.1 | NP_072124.1 | 297 | |||
GenBank mRNA | AF057026.1 | - | 297 | |||
AF086624.1 | - | 499 | ||||
BC097317.1 | - | 276 | 7446368 |