Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022634.2 | NP_072156.2 | 5 | 277 | ||
XM_006256078.2 | XP_006256140.1 | 5 | 418 | |||
XM_006256079.2 | XP_006256141.1 | 5 | 418 | |||
XM_006256080.2 | XP_006256142.1 | 4 | 365 | |||
GenBank mRNA | AF208230.1 | - | 5 | 277 | ||
FQ222284.1 | - | 4 | 275 |