Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022680.1 | NP_073171.1 | 3 | 216 | ||
GenBank mRNA | S66545.1 | - | 269 |