Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022683.1 | NP_073174.1 | 4, 5 | 562 | ||
XM_006243090.2 | XP_006243152.1 | 4, 5 | 1356 | |||
GenBank mRNA | AF135115.1 | - | 4, 5 | 562 | ||
FQ227844.1 | - | 4, 5 | 845 |