Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022799.2 | NP_073636.1 | 3 | 326 | ||
XM_006249797.2 | XP_006249859.1 | 3 | 398 | |||
GenBank mRNA | AJ237669.1 | - | 2 | 151 | ||
BC078818.1 | - | 3 | 326 | 7128528 | ||
FQ228159.1 | - | 3 | 354 |