Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1lc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022867.2 | NP_074058.2 | 3 | 255 | ||
GenBank mRNA | AY206669.1 | - | 3 | 217 | ||
AY310156.1 | - | 783 | ||||
AY392036.1 | - | 3 | 255 | |||
BC058144.1 | - | 3 | 213 | 6889616 | ||
BC083556.1 | - | 3 | 235 | 7189855 | ||
FQ221245.1 | - | 3 | 266 | |||
FQ228912.1 | - | 3 | 266 | |||
U05784.1 | - | 3 | 205 |