Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022934.1 | NP_075223.1 | 6 | 834 | ||
XM_006238099.1 | XP_006238161.1 | 6 | 932 | |||
GenBank mRNA | BC062009.1 | - | 6 | 791 | 5621557 | |
FQ216722.1 | - | 6 | 823 | |||
U53922.1 | - | 6 | 834 |