Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Doc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022937.2 | NP_075226.1 | 8 | 988 | ||
XM_006230335.2 | XP_006230397.1 | 9 | 1088 | |||
XM_006230336.1 | XP_006230398.1 | 9 | 1272 | |||
XM_006230337.2 | XP_006230399.1 | 9 | 1084 | |||
XM_006230339.2 | XP_006230401.1 | 9 | 1022 | |||
XM_006230340.1 | XP_006230402.1 | 9 | 1206 | |||
XM_006230341.2 | XP_006230403.1 | 9 | 1206 | |||
XM_008759918.1 | XP_008758140.1 | 9 | 1119 |