Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023971.1 | NP_076461.1 | 4 | 460 | ||
XM_006257235.2 | XP_006257297.1 | 3 | 431 | |||
XM_008773432.1 | XP_008771654.1 | 4 | 587 | |||
XM_008773433.1 | XP_008771655.1 | 4 | 343 | |||
XM_008773434.1 | XP_008771656.1 | 4 | 558 | |||
XM_008773435.1 | XP_008771657.1 | 3 | 431 | |||
XM_008773436.1 | XP_008771658.1 | 2 | 1896 | |||
GenBank mRNA | AF195787.1 | - | 4 | 460 | ||
AF195788.1 | - | 3 | 393 |