Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023977.2 | NP_076467.2 | 4 | 596 | ||
GenBank mRNA | AF311954.1 | - | 4 | 487 | ||
BC092568.1 | - | 4 | 596 | 7307277 |