Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npffr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023980.1 | NP_076470.1 | 2 | 352 | ||
XM_006250787.2 | XP_006250849.1 | 3 | 913 | |||
GenBank mRNA | AF268900.1 | - | 2 | 352 |
Set Membership: |
G-protein coupled receptor |