Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000795.3 | NP_000786.1 | 8 | 1938 | ||
NM_016574.3 | NP_057658.2 | 7 | 1851 | |||
XM_017017296.1 | XP_016872785.1 | 8 | 1920 | |||
GenBank mRNA | BC021195.2 | - | 8 | 1798 | 3939741 | |
M29066.1 | - | 7 | 1736 | |||
S69899.1 | - | 7 | 1713 | |||
X51362.1 | - | 8 | 1862 |
Set Membership: |
G-protein coupled receptor |