Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029896.1 | NP_001025067.1 | 9 | 954 | ||
NM_007075.3 | NP_009006.2 | 10 | 1224 | |||
GenBank mRNA | AJ005897.1 | - | 9 | 910 | ||
AK023344.1 | - | 7 | 1932 | |||
AK225122.1 | - | 9 | 933 | |||
AK315717.1 | - | 9 | 941 | |||
AY691428.1 | - | 8 | 782 | |||
BC000464.2 | - | 9 | 948 | 2822742 | ||
BC003037.1 | - | 9 | 948 | 2822742 | ||
BC009027.1 | - | 6 | 611 | |||
BC069206.1 | - | 10 | 1224 | 5801768 | ||
CR456725.1 | - | 8 | 782 |