Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030835.2 | NP_110462.1 | 3 | 779 | ||
GenBank mRNA | AB018546.1 | - | 3 | 664 | ||
AF100470.1 | - | 3 | 434 | |||
AJ238236.1 | - | 3 | 439 | |||
BC061854.1 | - | 3 | 779 | 5621665 |