Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc21a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030837.1 | NP_110464.1 | 11 | 1325 | ||
GenBank mRNA | D79981.1 | - | 11 | 1325 |