Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111098.1 | NP_001104568.1 | 5 | 479 | ||
NM_030857.2 | NP_110484.1 | 5 | 542 | |||
XM_006237834.2 | XP_006237896.1 | 5 | 858 | |||
GenBank mRNA | AF000300.1 | - | 5 | 387 | ||
AF000301.1 | - | 5 | 479 | |||
AF000302.1 | - | 5 | 477 | |||
L14782.1 | - | 4 | 336 | |||
L14823.1 | - | 4 | 273 | |||
L14951.1 | - | 5 | 545 |