Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007085.4 | NP_009016.1 | 11 | 1754 | ||
GenBank mRNA | AF288537.1 | - | 5 | 1034 | ||
BC000055.2 | - | 11 | 1661 | 3505833 | ||
BX647421.1 | - | 11 | 1679 | |||
U06863.1 | - | 11 | 1670 |