Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031043.2 | NP_112305.1 | 2 | 230 | ||
GenBank mRNA | AF021343.1 | - | 2 | 233 | ||
BC070944.1 | - | 3 | 271 | 7103004 | ||
L01793.1 | - | 1 | 50 |