Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000798.4 | NP_000789.1 | 1 | 2214 | ||
GenBank mRNA | BC009748.2 | - | 1 | 1869 | 3928370 |
Set Membership: |
G-protein coupled receptor |