Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031122.1 | NP_112384.1 | 12 | 1505 | ||
GenBank mRNA | BC078804.1 | - | 12 | 1512 | 7109473 | |
FQ212434.1 | - | 1562 | ||||
FQ228746.1 | - | 12 | 1475 | |||
X82021.1 | - | 12 | 1505 |