Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031154.1 | NP_112416.1 | 8 | 694 | ||
GenBank mRNA | BC059130.1 | - | 8 | 752 | 6921477 | |
FQ214048.1 | - | 8 | 743 | |||
J02744.1 | - | 8 | 694 |