Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031237.1 | NP_112516.1 | 7 | 905 | ||
XM_006233333.1 | XP_006233395.1 | 9 | 947 | |||
XM_006233334.1 | XP_006233396.1 | 9 | 1094 | |||
XM_006233335.1 | XP_006233397.1 | 9 | 901 | |||
XM_006233336.2 | XP_006233398.1 | 8 | 897 | |||
XM_006233337.1 | XP_006233399.1 | 8 | 1044 | |||
XM_006233338.1 | XP_006233400.1 | 8 | 882 | |||
GenBank mRNA | AB006852.1 | - | 7 | 905 | ||
AY724500.1 | - | 8 | 852 | |||
BC072696.1 | - | 8 | 828 | 7105617 | ||
FQ215509.1 | - | 4 | 482 | |||
FQ226278.1 | - | 8 | 881 | |||
FQ233355.1 | - | 8 | 872 | |||
U13175.1 | - | 7 | 759 |