Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007106.3 | NP_009037.1 | 5 | 4097 | ||
GenBank mRNA | AL080177.1 | - | 5 | 3065 | ||
BC044582.1 | - | 5 | 3543 | 5262176 | ||
BC059385.1 | - | 5 | 3441 | 5287541 |