Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252607.1 | NP_001239536.1 | 12 | 1559 | ||
NM_001252608.1 | NP_001239537.1 | 11 | 1226 | |||
NM_007112.4 | NP_009043.1 | 13 | 1586 | |||
NR_045553.1 | 12 | 1570 | ||||
XM_006711498.3 | XP_006711561.1 | 13 | 3145 | |||
XM_011509926.2 | XP_011508228.1 | 13 | 3145 | |||
XM_011509927.2 | XP_011508229.1 | 12 | 3118 | |||
XM_011509928.2 | XP_011508230.1 | 13 | 3052 | |||
XM_011509929.1 | XP_011508231.1 | 13 | 1541 | |||
XM_011509930.2 | XP_011508232.1 | 13 | 3145 | |||
XM_011509931.1 | XP_011508233.1 | 13 | 1491 | |||
XM_011509932.2 | XP_011508234.1 | 13 | 1424 | |||
XM_011509933.1 | XP_011508235.1 | 14 | 1773 | |||
XM_011509934.1 | XP_011508236.1 | 13 | 1528 | |||
XM_011509935.1 | XP_011508237.1 | 13 | 1684 | |||
XM_011509936.1 | XP_011508238.1 | 12 | 1425 | |||
XM_017002206.1 | XP_016857695.1 | 12 | 1498 | |||
GenBank mRNA | AK074796.1 | - | 10 | 922 | ||
AK298592.1 | - | 11 | 1162 | |||
AK298803.1 | - | 10 | 1054 | |||
AK298914.1 | - | 3 | 236 | |||
AK314337.1 | - | 12 | 1510 | |||
BC018786.1 | - | 4 | 389 | |||
BC113846.1 | - | 12 | 1488 | 40008346 | ||
BC113847.1 | - | 13 | 1504 | 40008343 | ||
L38969.1 | - | 13 | 1502 |