Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031811.3 | NP_113999.2 | 1191 | |||
GenBank mRNA | BC059126.1 | - | 1195 | 6889440 | ||
FQ219168.1 | - | 1217 |