Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031839.3 | NP_114027.2 | 9 | 1623 | ||
GenBank mRNA | BC078707.1 | - | 9 | 1623 | 7110580 | |
FQ209508.1 | - | 7 | 1222 | |||
FQ218733.1 | - | 4 | 750 | |||
FQ218882.1 | - | 6 | 971 | |||
U04733.1 | - | 9 | 1604 |