Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007120.2 | NP_009051.1 | 1 | 558 | ||
GenBank mRNA | AK313623.1 | - | 1 | 572 | ||
AY435139.1 | - | 1 | 529 | |||
BC121036.1 | - | 1 | 529 | 40117585 | ||
BC131623.1 | - | 1 | 522 | |||
BC139784.1 | - | 1 | 549 | 40117089 | ||
DQ364249.1 | - | 1 | 538 | |||
M57951.1 | - | 1 | 558 |