Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032066.1 | NP_114455.1 | 5, 6 | 475 | ||
XM_006234599.2 | XP_006234661.1 | 5, 6 | 642 | |||
GenBank mRNA | BC061543.1 | - | 5, 6 | 496 | 6888949 | |
FQ213557.1 | - | 5, 6 | 511 | |||
FQ214777.1 | - | 5, 6 | 540 | |||
U81186.1 | - | 5, 6 | 475 |