Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053374.2 | NP_445826.2 | 3 | 326 | ||
XM_008759805.1 | XP_008758027.1 | 3 | 2484 | |||
GenBank mRNA | AF154569.1 | - | 2 | 137 | ||
CB809932.1 | - | 3 | 338 |