Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053389.1 | NP_445841.1 | 3 | 366 | ||
GenBank mRNA | AF176072.1 | - | 3 | 366 | ||
BC098662.1 | - | 3 | 309 | 7386123 |