Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007148.4 | NP_009079.2 | 11, 12 | 1526 | ||
XM_006721571.3 | XP_006721634.1 | 11, 12 | 5558 | |||
XM_006721572.3 | XP_006721635.1 | 11, 12 | 5556 | |||
XM_006721573.3 | XP_006721636.1 | 11, 12 | 1291 | |||
GenBank mRNA | AB026054.1 | - | 11, 12 | 1383 | ||
AK226052.1 | - | 11, 12 | 1466 | |||
AK313844.1 | - | 11, 12 | 1472 | |||
BC053989.1 | - | 11, 12 | 1441 | 5922474 |