Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053427.1 | NP_445879.1 | 4 | 987 | ||
XM_006229265.1 | XP_006229327.1 | 4 | 516 | |||
GenBank mRNA | AF271235.1 | - | 4 | 987 |