Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF195 SIRNA Assays ›
Gene Symbol:
ZNF195
Gene Name:
zinc finger protein 195
Gene Aliases:
HRF1, ZNFP104
Chromosome Location:
Chr.11: 3357927 - 3379222 on Build GRCh38
UniGene ID:
Hs.386294
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19276
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF195
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130519.2
NP_001123991.1
5
678
NM_001130520.2
NP_001123992.1
6
747
NM_001242841.1
NP_001229770.1
6
758
NM_001242842.1
NP_001229771.1
6
680
NM_001242843.1
NP_001229772.1
5
611
NM_001256823.1
NP_001243752.1
6
668
NM_001256824.1
NP_001243753.1
6
704
NM_001256825.1
NP_001243754.1
5
630
NM_007152.4
NP_009083.2
4
531
NR_040083.1
6
895
NR_046381.1
7
815
NR_046382.1
5
717
XM_006718306.1
XP_006718369.1
5
734
XM_011520350.2
XP_011518652.1
7
736
XM_011520351.2
XP_011518653.1
8
1107
XM_011520352.2
XP_011518654.1
6
803
XM_011520354.2
XP_011518656.2
5
766
XM_017018260.1
XP_016873749.1
7
950
XM_017018261.1
XP_016873750.1
7
832
XM_017018262.1
XP_016873751.1
6
881
XM_017018263.1
XP_016873752.1
6
696
XM_017018264.1
XP_016873753.1
4
548
XM_017018265.1
XP_016873754.1
7
811
XM_017018266.1
XP_016873755.1
6
731
XM_017018267.1
XP_016873756.1
8
1084
GenBank mRNA
AB209664.1
-
1
8664
AF003540.1
-
6
610
AK095720.1
-
4
454
AK131296.1
-
6
814
AK290099.1
-
6
656
AK293679.1
-
6
680
AK308024.1
-
6
692
AK308068.1
-
6
597
AK308209.1
-
5
572
AL833722.1
-
4
444
BC121028.1
-
4
387
40117443
BC121029.1
-
5
534
40117445
BM462194.1
-
6
573
CX867141.1
-
5
574
DA618142.1
-
5
547
DA765121.1
-
2
236
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline