Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053500.1 | NP_445952.1 | 7, 8 | 855 | ||
XM_006244610.2 | XP_006244672.1 | 7, 8 | 971 | |||
XM_006244611.2 | XP_006244673.1 | 7, 8 | 971 | |||
XM_006244612.2 | XP_006244674.1 | 7, 8 | 971 | |||
XM_008766882.1 | XP_008765104.1 | 7, 8 | 1191 | |||
GenBank mRNA | AJ300162.1 | - | 7, 8 | 855 | ||
AJ300163.1 | - | 7, 8 | 942 | |||
AJ300164.1 | - | 7, 8 | 963 | |||
AY512661.1 | - | 7, 8 | 785 |