Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053514.1 | NP_445966.1 | 602 | |||
XM_006241305.2 | XP_006241367.1 | 4 | 505 | |||
GenBank mRNA | AF090135.1 | - | 602 |