Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000839.3 | NP_000830.2 | 3 | 714 | ||
XM_011533635.1 | XP_011531937.1 | 2 | 2385 | |||
XM_011533637.1 | XP_011531939.1 | 2 | 710 | |||
XM_011533638.2 | XP_011531940.1 | 3 | 452 | |||
XM_011533639.2 | XP_011531941.1 | 3 | 170 | |||
XM_011533640.2 | XP_011531942.1 | 3 | 123 | |||
XM_011533641.2 | XP_011531943.1 | 3 | 159 | |||
XM_011533642.1 | XP_011531944.1 | 2 | 2385 | |||
XM_017006271.1 | XP_016861760.1 | 3 | 1192 | |||
GenBank mRNA | AB209164.1 | - | 3 | 1204 | ||
AK289984.1 | - | 3 | 707 | |||
AK302671.1 | - | 2 | 473 | |||
AK313056.1 | - | 3 | 691 | |||
AK315816.1 | - | 3 | 709 | |||
AY999299.1 | - | 2 | 511 | |||
BC113615.1 | - | 2 | 498 | 8322667 | ||
BC113619.1 | - | 2 | 498 | 8322671 | ||
BX648951.1 | - | 2 | 2154 | |||
EU432122.1 | - | 2 | 475 | |||
L35318.1 | - | 2 | 477 |
Set Membership: |
G-protein coupled receptor |