Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053583.2 | NP_446035.2 | 4081 | |||
GenBank mRNA | L03386.1 | - | 3446 | |||
U92564.1 | - | 4079 |