Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053746.1 | NP_446198.1 | 21, 22 | 3059 | ||
XM_008765902.1 | XP_008764124.1 | 21, 22 | 3124 | |||
GenBank mRNA | D87212.1 | - | 21, 22 | 3059 |