Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053770.1 | NP_446222.1 | 5, 6 | 818 | ||
GenBank mRNA | AF026505.1 | - | 5, 6 | 818 |