Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NISCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007184.3 | NP_009115.2 | 21 | 4715 | ||
XM_005264839.2 | XP_005264896.1 | 20 | 4564 | |||
XM_006712955.2 | XP_006713018.1 | 9 | 5242 | |||
XM_017005643.1 | XP_016861132.1 | 12 | 3556 | |||
GenBank mRNA | AB023192.1 | - | 22 | 4653 | ||
AF082516.1 | - | 21 | 4607 | |||
AK001505.1 | - | 18 | 4830 | |||
AK027331.1 | - | 7 | 2526 | |||
AK027428.1 | - | 21 | 4593 | |||
AK097732.1 | - | 5 | 1678 | |||
AK297657.1 | - | 7 | 1391 | |||
AK302441.1 | - | 12 | 3603 | |||
AL117432.1 | - | 8 | 3174 | |||
BC038102.2 | - | 21 | 4619 | 5268247 | ||
BC054494.1 | - | 21 | 4613 | 6195532 | ||
BC056900.1 | - | 21 | 4585 | 6143316 |