Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gripap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053807.1 | NP_446259.1 | 26 | 2558 | ||
GenBank mRNA | AF274057.1 | - | 26 | 2558 | ||
AF274058.1 | - | 24 | 2423 | |||
FQ233233.1 | - | 5 | 557 |