Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053818.2 | NP_446270.1 | 5, 6 | 788 | ||
XM_006238625.2 | XP_006238687.1 | 4, 5 | 794 | |||
GenBank mRNA | BC128732.1 | - | 5, 6 | 788 | 8364347 | |
L13600.1 | - | 4, 5 | 685 | |||
M88595.1 | - | 4, 5 | 676 | |||
M95413.1 | - | 5, 6 | 739 |