Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053859.2 | NP_446311.1 | 4 | 655 | ||
GenBank mRNA | EU253553.1 | - | 5 | 620 | ||
FQ211811.1 | - | 668 | ||||
U07609.1 | - | 4 | 658 |