Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007198.3 | NP_009129.1 | 3 | 298 | ||
XM_017013001.1 | XP_016868490.1 | 3 | 1128 | |||
GenBank mRNA | AK021923.1 | - | 3 | 166 | ||
AL136616.1 | - | 3 | 278 | |||
BC012334.1 | - | 3 | 288 | 3546307 | ||
CR533532.1 | - | 3 | 221 | |||
DA086552.1 | - | 3 | 298 |