Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053911.2 | NP_446363.1 | 2 | 308 | ||
XM_006228964.2 | XP_006229026.1 | 2 | 326 | |||
XM_008759304.1 | XP_008757526.1 | 1 | 799 | |||
GenBank mRNA | FQ229744.1 | - | 1 | 29 | ||
U83896.1 | - | 2 | 312 |