Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053928.2 | NP_446380.1 | 2 | 420 | ||
GenBank mRNA | AB032739.1 | - | 2 | 81 | ||
BC090072.1 | - | 2 | 420 | 7113593 |