Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053968.3 | NP_446420.1 | 3 | 286 | ||
GenBank mRNA | BC058453.1 | - | 3 | 289 | 6887014 | |
S65838.1 | - | 3 | 284 |