Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RragB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053972.2 | NP_446424.1 | 3 | 736 | ||
XM_006256779.2 | XP_006256841.1 | 3 | 834 | |||
XM_008773066.1 | XP_008771288.1 | 2 | 891 | |||
XM_008773067.1 | XP_008771289.1 | 2 | 891 | |||
GenBank mRNA | BC078760.1 | - | 3 | 274 | 7123902 | |
BC087698.1 | - | 3 | 736 | 7314028 |
Gene Symbol: LOC100909655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752022.3 | XP_003752070.1 | 835 | |||
XM_006256775.2 | XP_006256837.1 | 834 | ||||
XM_008773107.1 | XP_008771329.1 | 891 | ||||
XM_008773108.1 | XP_008771330.1 | 891 | ||||
XM_008773109.1 | XP_008771331.1 | 2 | 891 |