Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RragB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053972.2 | NP_446424.1 | 4 | 797 | ||
XM_008773066.1 | XP_008771288.1 | 3 | 952 | |||
GenBank mRNA | BC078760.1 | - | 4 | 335 | 7123902 | |
BC087698.1 | - | 4 | 797 | 7314028 |
Gene Symbol: LOC100909655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752022.3 | XP_003752070.1 | 896 | |||
XM_008773107.1 | XP_008771329.1 | 952 | ||||
XM_008773109.1 | XP_008771331.1 | 3 | 952 |